Learn More
Med Vet International Jorvet Thermoblocker Recovery bag, 115cm x 60cm, J0955, Each
Catalog No. 5000495251
Encompass_Preferred
The Jorvet 115cm Thermoblocker Recovery Bag, has a thin silver reflective coating to back radiant heat. It is utilized with search and rescue teams to help animals concerve heat. With this recovery bag almost no heat passes throught the Thermoblocker. This product is easy to clean, is lightweight, and has padding that adds comfort for the patient. Features: The top reflective layer returns nearly 80% of the radiant heat Any heat that passes through is slowed by insulating bubble wrap and kept in the material by the second reflective layer
Catalog No. 50-004-95251 Supplier Med Vet International Supplier No. JOR955
The Jorvet 115cm Thermoblocker Recovery Bag, has a thin silver reflective coating to back radiant heat. It is utilized with search and rescue teams to help animals concerve heat. With this recovery bag almost no heat passes throught the Thermoblocker. This product is easy to clean, is lightweight, and has padding that adds comfort for the patient. Features: The top reflective layer returns nearly 80% of the radiant heat Any heat that passes through is slowed by insulating bubble wrap and kept in the material by the second reflective layer