Learn More
Hach Company Stand-Up Sterile Bag with Sodium Thiosulfate, pk/100
Supplier:  Hach Company 2075533
Encompass
Specially designed for chlorinated water samples (complies with USEPA and Standard Methods Criteria). Sterile (certified), disposable, transparent polyethylene bags with trusted "Whirl-Pak" closure. Contains non-nutritive pill with 10 mg sodium thiosulfate.
Catalog No. 50-199-5235