Learn More
Hach Company Bag, Sterile, Whirl-Pak with Dechlorinating Agent, 100 mL, 100/pk
Catalog No. 501995232 Shop All Hach Company Products
Encompass
Specially designed for chlorinated water samples (Complies with USEPA and Standard Methods criteria). Sterile (certified), disposable. transparent polyethylene bags with trusted “Whirl-Pak” closure. Contains non-nutritive pill with 10 mg sodium thiosulfate (will dechlorinate up to 10 mg/Cl2 in 100 mL sample ) Four ounce and 100 mL fill-lines. “Write-on” strip accepts waterproof ink.
Catalog No. 50-199-5232 Supplier Hach Company Supplier No. 2075333
Specially designed for chlorinated water samples (Complies with USEPA and Standard Methods criteria). Sterile (certified), disposable. transparent polyethylene bags with trusted “Whirl-Pak” closure. Contains non-nutritive pill with 10 mg sodium thiosulfate (will dechlorinate up to 10 mg/Cl2 in 100 mL sample ) Four ounce and 100 mL fill-lines. “Write-on” strip accepts waterproof ink.