Learn More
Hach Company Bag-Sterile-WhirlPak-720 mL
Supplier:  Hach Company 1437297
Encompass
Bag-Sterile-WhirlPak-720 mL-10/pkComplies with Standard Methods and USEPA for drinking water sample collection Made from poly resins approved by USDA and FDA for food contact Sterile (certified) disposable transparent polyethylene bags with trusted in Whirl-Pakin closure Suitable for liquid or solid sample collection in Write-onin surface accepts waterproof ink 24 oz bag perfect for incubating petri dishes in water bath applications
Catalog No. 50-002-57154