Learn More
Hach Company Bag, Sterile, WhirlPak, 207 mL, 500/pk
Supplier:  Hach Company 2233100
Encompass
Approved by U.S. Environmental Protection Agency (USEPA). Disposable, transparent polyethylene bags. Gas-sterilized, pull-tab openings. Can be marked with waterproof felt pen or marker. Sure "whirl" closure. Nasco.
Catalog No. 50-199-5586