Learn More
Essendant High-Density Commercial Can Liners, 60 gal, 22 microns, 43"" x 48"", Black, 150/Carton
Catalog No. NC0836466
Encompass_Preferred
Made from high-quality virgin resins for reliability and consistency. Star-sealed bottom distributes weight evenly and creates a seal up to eight times stronger than the film itself. Innovative "clamshell" box with E-Z identification eliminates confusion. ISO registered facilities ensure superior quality and consistency.
Catalog No. NC0836466 Supplier Essendant Supplier No. IBSS434822K
Made from high-quality virgin resins for reliability and consistency. Star-sealed bottom distributes weight evenly and creates a seal up to eight times stronger than the film itself. Innovative "clamshell" box with E-Z identification eliminates confusion. ISO registered facilities ensure superior quality and consistency.