Learn More
Elkay Plastics Company Bag Specimen Transport 3 wall Yellow Tint with Removable Biohazard logo and Outside Pouch, Reclosable Closure 1.75 mil 6 x 9 1,000 per Case
Catalog No. 501049763
Encompass_Preferred
LKs Lab-Loc Reclosable Zipper Specimen Bags with Seal-N-Rip removable biohazard symbols provide an excellent means for securely transferring specimens in hospital and medical environments while helping to save on disposal costs. The bags have a special removable biohazard symbol which can be torn off and allows the bags to be disposed of with regular waste if they are uncontaminated.
Catalog No. 50-104-9763 Supplier Elkay Plastics Company Supplier No. LABZ69YE
LKs Lab-Loc Reclosable Zipper Specimen Bags with Seal-N-Rip removable biohazard symbols provide an excellent means for securely transferring specimens in hospital and medical environments while helping to save on disposal costs. The bags have a special removable biohazard symbol which can be torn off and allows the bags to be disposed of with regular waste if they are uncontaminated.