Learn More
Elkay Plastics Company Bag Specimen Transport Zip Closure 3 wall Assorted with Biohazard print and an Outside Pouch for documents 2 mil 6 X 9 1,000 per Case
Catalog No. NC2743894
Encompass_Preferred
LKs Lab-Loc Reclosable Zipper Specimen Bags provide an excellent means for transferring specimens in hospital and medical environments. Three wall construction provides a handy outer pouch to contain accompanying paperwork during specimen transfer. Bags are inner packed in 100s.
Catalog No. NC2743894 Supplier Elkay Plastics Company Supplier No. LAB20609AS
LKs Lab-Loc Reclosable Zipper Specimen Bags provide an excellent means for transferring specimens in hospital and medical environments. Three wall construction provides a handy outer pouch to contain accompanying paperwork during specimen transfer. Bags are inner packed in 100s.