Learn More
Elkay Plastics Company Bag Specimen Transport 3 wall Clear with Removable Biohazard logo and Outside Pouch, Reclosable Closure 2 mil 14 X 20 250 per Case
Catalog No. NC2743960
Encompass_Preferred
LKs Lab-Loc Reclosable Zipper Specimen Bags with Seal-N-Rip feature provide an excellent means for securely transferring specimens in hospital and medical environments. Their three wall construction provides a handy outer pouch with flaps to contain and secure accompanying paperwork during specimen transfer
Catalog No. NC2743960 Supplier Elkay Plastics Company Supplier No. LABZ1420B
LKs Lab-Loc Reclosable Zipper Specimen Bags with Seal-N-Rip feature provide an excellent means for securely transferring specimens in hospital and medical environments. Their three wall construction provides a handy outer pouch with flaps to contain and secure accompanying paperwork during specimen transfer