Learn More
Elkay Plastics Company Bag Specimen Transport 3 wall Yellow Tint with Removable Biohazard logo and Outside Pouch, Reclosable Closure 1.75 mil 8 x 10 1,000 per Case
Catalog No. NC2743991
Encompass_Preferred
LKs Lab-Loc Reclosable Zipper Specimen Bags with Seal-N-Rip removable biohazard symbols provide an excellent means for securely transferring specimens in hospital and medical environments while helping to save on disposal costs. The bags have a special removable biohazard symbol which can be torn off and allows the bags to be disposed of with regular waste if they are uncontaminated.
Catalog No. NC2743991 Supplier Elkay Plastics Company Supplier No. LABZ810B
LKs Lab-Loc Reclosable Zipper Specimen Bags with Seal-N-Rip removable biohazard symbols provide an excellent means for securely transferring specimens in hospital and medical environments while helping to save on disposal costs. The bags have a special removable biohazard symbol which can be torn off and allows the bags to be disposed of with regular waste if they are uncontaminated.