Learn More
Aviva Systems Biology Corp BAG1 Antibody - N-terminal region (ARP63477_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63477_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BAG1. It was validated on Western Blot. . Alternate names: HAP, RAP46ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Zebrafish
Catalog No. 50-287-982