Learn More
Aviva Systems Biology Corp BAG2 Antibody - C-terminal region (ARP42177_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42177_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against BAG2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human BAG2. Alternate names: RP3-496N17.2, BAG-2, KIAA0576, MGC149462, dJ417I1.2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58132