Learn More
Aviva Systems Biology Corp BAG4 Antibody - C-terminal region (ARP61938_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61938_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BAG4. It was validated on Western Blot. . Alternate names: BAG-4, SODDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-287-511