Learn More
Aviva Systems Biology Corp BAG5 Antibody - N-terminal region (ARP61997_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61997_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BAG5. It was validated on Western Blot. . Alternate names: BAG-5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-287-538