Learn More
Interscience BagOpen Jumbo Kit
SDP
Catalog No. 16949069 Shop All Interscience Products
Change view
Click to view available options
For Use With (Equipment):
3500 mL Bags
Catalog No. For Use With (Equipment)
16-949-069 3500 mL Bags
1 options
Catalog No. 16-949-069 Supplier Interscience Supplier No. 505006
Add to Cart
Add to Cart
Jumbo kit for DiluFlow including BagOpen and drip tray.
Jumbo BagOpen kit for use with DiluFlow
Kit includes:
  • BagOpen 3500
  • 3,500 mL drip tray
Specifications
Product Type Bag Opener Kit
Autoclavable Not Autoclavable
For Use With (Equipment) 3500 mL Bags
Sterility Non-sterile