Learn More
MTC Bio Autoclave Bags, 200/PK
Catalog No. 501870972
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Autoclave bags, 12.2x26" (31 x 66cm), clear, biohazard, printed, marking area
Catalog No. 50-187-0972 Supplier MTC Bio Supplier No. A9000C
This item has a minimum qty of 2 per supplier requirements.
Autoclave bags, 12.2x26" (31 x 66cm), clear, biohazard, printed, marking area