Learn More
Med Vet International Specimen Biohazard Transport Bags, Glue Closure, 6" x 9", J1493, 100/Pack
Catalog No. 5000495535
Encompass_Preferred
Specimen Transport Bags - Glue Closure The special glue closure of our specimen transport bags offer a liquid-tight tamper evident seal. Simply pull off the liner to expose the glue adhesive and press to close. 6" x 9" Bag, 100 /pk
Catalog No. 50-004-95535 Supplier Med Vet International Supplier No. JOR1493
Specimen Transport Bags - Glue Closure The special glue closure of our specimen transport bags offer a liquid-tight tamper evident seal. Simply pull off the liner to expose the glue adhesive and press to close. 6" x 9" Bag, 100 /pk