Learn More
Med Vet International Specimen Biohazard Transport Bags w/ Absorbent Pad, Zipper Closure, 6" x 9", J1492B, 100/Pack
Catalog No. 5000495534
Encompass_Preferred
Specimen Transport Bags - Zipper Closure Designed for safe transport of specimens and documentation. Each bag is printed with Biohazard symbol and offers checkmark boxes to indicate special handling instructions. • 2 mil gauge polyethylne (PE) • Attached document compartment • Zipper closure allows repeated access to specimen • Imprinted biohazard symbol • Complies with OSHA and NCCLS guidelines •One option is absorbent pad that can hold 60 ml of body fluids 6" x 9" Bag w/ Absorbent Pad, 100 /pk
Catalog No. 50-004-95534 Supplier Med Vet International Supplier No. JOR1492B
Specimen Transport Bags - Zipper Closure Designed for safe transport of specimens and documentation. Each bag is printed with Biohazard symbol and offers checkmark boxes to indicate special handling instructions. • 2 mil gauge polyethylne (PE) • Attached document compartment • Zipper closure allows repeated access to specimen • Imprinted biohazard symbol • Complies with OSHA and NCCLS guidelines •One option is absorbent pad that can hold 60 ml of body fluids 6" x 9" Bag w/ Absorbent Pad, 100 /pk