Learn More
Novus Biologicals™ BAHCC1 Recombinant Protein Antigen

Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human BAHCC1
Source: E.coli
Amino Acid Sequence:
SGLDKSGYFELPTSSQDCARPGHQDPLGGKAPQACCTLDKTVGKEAPAGPPGAQKVARIRHQQHLMAAEVEQGGIGAEAKRKSLELA
Specifications
Specifications
| Gene ID (Entrez) | 57597 |
| Purity | >80% by SDS-PAGE and Coomassie blue staining |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Formulation | PBS and 1M Urea, pH 7.4. |
| For Use With (Application) | Antibody Competition |
| Gene Alias | BAH and coiled-coil domain-containing protein 1, BAH domain and coiled-coil containing 1, BAH domain-containing protein 2, BAHD2, bromo adjacent homology domain-containing protein 2, FLJ23058, KIAA1447 |
| Gene Symbol | BAHCC1 |
| Molecular Weight (g/mol) | 27 kDa |
| Quantity | 100 μL |
| Source | E. coli |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.