Learn More
Selleck Chemical LLC BAI1-5mg
SDP
Catalog No. 502389977
Encompass_Preferred
BAI1 is a direct allosteric inhibitor of BAX with a dissociation constant (Kd) of 15.0 ± 4 μM.
Catalog No. 50-238-9977 Supplier Selleck Chemical LLC Supplier No. S88655MG
BAI1 is a direct allosteric inhibitor of BAX with a dissociation constant (Kd) of 15.0 ± 4 μM.