Learn More
Medchemexpress LLC BAI3 protein, human (HEK293, His) | 96.6% | 100 UG
SDP
Catalog No. 5000293958
Encompass_Preferred
Recombinant human brain-specific angiogenesis inhibitor 3 (BAI3) produced in HEK293 cells with a C-terminal His tag. Purified protein intended for biochemical and cell biology research, particularly studies of adhesion GPCR function and interactions with C1q-like proteins. Provided in a 100 μg pack; consult supplier documentation for storage and handling.
  • Recombinant human protein expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • High purity (>95% by reducing SDS-PAGE).
  • Provided in 100 μg pack sizes.
  • Suitable for biochemical and cell biology assays.
Catalog No. 50-002-93958 Supplier Medchemexpress LLC Supplier No. HYP7675100UG
Recombinant human brain-specific angiogenesis inhibitor 3 (BAI3) produced in HEK293 cells with a C-terminal His tag. Purified protein intended for biochemical and cell biology research, particularly studies of adhesion GPCR function and interactions with C1q-like proteins. Provided in a 100 μg pack; consult supplier documentation for storage and handling.
  • Recombinant human protein expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • High purity (>95% by reducing SDS-PAGE).
  • Provided in 100 μg pack sizes.
  • Suitable for biochemical and cell biology assays.