Learn More
Abcam Baicalein, Lipo x ygenase inhibitor., 50MG
Catalog No. 502707335
Encompass_Preferred
MW 270.24 g/mol, Purity >98%. Cell-permeable flavonoid. Lipoxygenase inhibitor (IC₅₀ values are 0.64 and 1.6 μM at 12- and 15-lipoxygenases, respectively). Shows antioxidant and anti-inflammatory properties. Also induces apoptosis. Additionally, inhibits α-syn oligomerisation, β-Amyloid induced neurotoxicity and amyloid protein aggregation.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7335 Supplier Abcam Supplier No. AB12072350MG
MW 270.24 g/mol, Purity >98%. Cell-permeable flavonoid. Lipoxygenase inhibitor (IC₅₀ values are 0.64 and 1.6 μM at 12- and 15-lipoxygenases, respectively). Shows antioxidant and anti-inflammatory properties. Also induces apoptosis. Additionally, inhibits α-syn oligomerisation, β-Amyloid induced neurotoxicity and amyloid protein aggregation.
The product is subject to the following: Abcam Restricted Use Statement