Learn More
Medchemexpress LLC HY-N0197 100mg Medchemexpress, Baicalin CAS:21967-41-9 Purity:>98%
SDP
Catalog No. 501933460
Encompass_Preferred
Medchemexpress, HY-N0197 100mg Baicalin CAS:21967-41-9 Baicalin is a flavonoid glycoside isolated from Scutellaria baicalensis. Baicalin reduces the expression of NF-κB . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3460 Supplier Medchemexpress LLC Supplier No. HYN0197100MG
Medchemexpress, HY-N0197 100mg Baicalin CAS:21967-41-9 Baicalin is a flavonoid glycoside isolated from Scutellaria baicalensis. Baicalin reduces the expression of NF-κB . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.