Learn More
Cole-Parmer Polyethylene Bailer, 36" (91.4 cm) long, 0.75" (1.9 cm) OD, 1 Check Valve
Catalog No. NC1919989 Shop All Cole-Parmer Products
Encompass_Preferred
These disposable bailers feature tapered ends and low-snag cord cleat to reduce hang-ups and snags. The angled nozzle ensures positive check-valve control. Choose from virgin polyethylene construction for strength and durability, PVC for strength and economy of polyethylene with transparent features, or FEP construction for high-purity applications. Double check-valve bailers allow you to sample from a specific depth (point-source sampling).
Catalog No. NC1919989 Supplier Cole-Parmer Supplier No. 9915730
These disposable bailers feature tapered ends and low-snag cord cleat to reduce hang-ups and snags. The angled nozzle ensures positive check-valve control. Choose from virgin polyethylene construction for strength and durability, PVC for strength and economy of polyethylene with transparent features, or FEP construction for high-purity applications. Double check-valve bailers allow you to sample from a specific depth (point-source sampling).