Learn More
Cole-Parmer Bain Marie Storage Container, HDPE, 20 qt; 6/PK
Supplier:  Cole-Parmer 6353012
Encompass_Preferred
Containers are made of FDA-compliant HDPE and can be frozen, boiled, or chemically sterilized. Snap-tight, positive-lock lids are sold separately.
Catalog No. NC4001329