Learn More
Alkali Scientific Baird Parker Agar, 1 Kg
SDP
Catalog No. 5000660439
Encompass_Preferred
Baird Parker Agar is a selective medium designed for the growth of coagulase-positive staphylococci in laboratory environments. Available in a 1kg pack, it is used primarily for the isolation of pathogenic bacteria, such as Staphylococcus aureus, from food and clinical samples. This agar medium supports accurate microbial testing and is essential for food safety and clinical microbiology labs.
Catalog No. 50-006-60439 Supplier Alkali Scientific Supplier No. AIR001CP
Baird Parker Agar is a selective medium designed for the growth of coagulase-positive staphylococci in laboratory environments. Available in a 1kg pack, it is used primarily for the isolation of pathogenic bacteria, such as Staphylococcus aureus, from food and clinical samples. This agar medium supports accurate microbial testing and is essential for food safety and clinical microbiology labs.