Learn More
Bioworld Baird Parker Agar, 2 kg
Catalog No. 50254273
Encompass_Preferred
Baird Parker AgarCommonly used in the detection and enumeration of coagulase-positive staphylococci in food samplesContains potassium tellurite and lithium which supresses the growth of most bacteria but allows the growth of staphylococcus aureuspH: 6.9Dehydrated Appearance: light tan, homogeneous powderPrepared Appearance: yellow agarSoluble in boiling waterContains:Pancreatic Digest of Casein Beef Extractive Yeast Extract Lithium Chloride Glycine Sodium Pyruvate Agar
Catalog No. 50-254-273 Supplier Bioworld Supplier No. 306200063
Baird Parker AgarCommonly used in the detection and enumeration of coagulase-positive staphylococci in food samplesContains potassium tellurite and lithium which supresses the growth of most bacteria but allows the growth of staphylococcus aureuspH: 6.9Dehydrated Appearance: light tan, homogeneous powderPrepared Appearance: yellow agarSoluble in boiling waterContains:Pancreatic Digest of Casein Beef Extractive Yeast Extract Lithium Chloride Glycine Sodium Pyruvate Agar