Learn More
NEOGEN Baird Parker Agar Base, 5 kg
SDP
Catalog No. 502015718
Encompass_Preferred
Baird Parker Agar is used for the cultivation of Staphylococcus aureus from foodstuffs according to ISO 6888, and is not intended for use in the diagnosis of disease or other conditions in humans.Originally introduced in 1962, this medium was developed by Baird-Parker to overcome the problems of recovering damaged Staphylococcus aureus from foodstuffs. This version of the medium is formulated according to ISO 6888-1:1999+A1:2003 and is in compliance with ISO 6888-2:2003+A1:2003 and ISO 6888-3:2003.Baird-Parker medium is highly selective by nature, due to the presence of potassium tellurite and lithium chloride. Tellurite inhibits most coliforms and is also reduced to telluride by S. aureus, giving the typical black colonies. Glycine and sodium pyruvate are both used as growth factors by staphylococci while the pyruvate also neutralizes any toxic peroxides that may be formed.For more information visit https://www.neogen.com/solutions/Microbiology/baird-parker-agar-base
Catalog No. 50-201-5718 Supplier NEOGEN Supplier No. 700004639
Baird Parker Agar is used for the cultivation of Staphylococcus aureus from foodstuffs according to ISO 6888, and is not intended for use in the diagnosis of disease or other conditions in humans.Originally introduced in 1962, this medium was developed by Baird-Parker to overcome the problems of recovering damaged Staphylococcus aureus from foodstuffs. This version of the medium is formulated according to ISO 6888-1:1999+A1:2003 and is in compliance with ISO 6888-2:2003+A1:2003 and ISO 6888-3:2003.Baird-Parker medium is highly selective by nature, due to the presence of potassium tellurite and lithium chloride. Tellurite inhibits most coliforms and is also reduced to telluride by S. aureus, giving the typical black colonies. Glycine and sodium pyruvate are both used as growth factors by staphylococci while the pyruvate also neutralizes any toxic peroxides that may be formed.For more information visit https://www.neogen.com/solutions/Microbiology/baird-parker-agar-base