Learn More
MSC Syngenta Crop Protection Ant Bait Arena, 12 x 1.98 gm, Indoxacarb
Catalog No. 502836044
Encompass
This ant bait arena provides targeted, thorough control of nuisance ants. It features the MetaActive™ effect and contains a highly attractive formulation. The bait is encapsulated in small plastic arenas for easy placement where ants are foraging.
  • Contains Indoxacarb as the active ingredient.
  • Comes in a quantity of 12 x 1.98 gm units.
  • Designed as a bait station.
  • Effective against ants.
  • Suitable for commercial, pest management, and residential pest control sites.
Catalog No. 50-283-6044 Supplier MSC Supplier No. 74323668
This ant bait arena provides targeted, thorough control of nuisance ants. It features the MetaActive™ effect and contains a highly attractive formulation. The bait is encapsulated in small plastic arenas for easy placement where ants are foraging.
  • Contains Indoxacarb as the active ingredient.
  • Comes in a quantity of 12 x 1.98 gm units.
  • Designed as a bait station.
  • Effective against ants.
  • Suitable for commercial, pest management, and residential pest control sites.