Learn More
Enzo Life Sciences Bak (human) monoclonal antibody (TC100) (50 µg)
SDP
Supplier:  Enzo Life Sciences BMLSA2980050
Encompass_Preferred
Clone: TC100. Isotype: IgG2a. Immunogen: Recombinant N-terminal human bak. Species reactivity: Human. Applications: Flow Cytometry, ICC, WB. Purity: Purified. Formulation: Liquid. In PBS containing sodium azide and gelatin.
Catalog No. 50-201-1796