Learn More
Enzo Life Sciences Bak polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADIAAP030F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human Bak. UniProt ID: Q16611. Source: Purified from rabbit serum. Species reactivity: Human Porcine, Monkey, Hamster. Applications: IF, IHC, IP, WB. Recommended Dilutions: Immunohistochemistry (1:50) Immunoprecipitation(1:100) Western Blot (1:1,000, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7149