Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Baker-flex™, Silica Gel IB2, J.T. Baker™

Catalog No. 02003905 Shop All Avantor Products
Change view
Click to view available options
Dimensions (L x W):
2.5 x 7.5 cm
5 x 20 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
02-003-905 5 x 20 cm
02-003-906 2.5 x 7.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 02-003-905 Supplier Avantor Supplier No. 444800
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Flexible TLC Sheet

  • Material: High-purity silica gel IB2 coating with inert binder
  • Substrate: Lightweight, unbreakable, and flexible plastic
  • Performance: Abrasion-resistant, uniform coating; resists attack by reagents and solvents
  • Chemical Properties: Inert binder and indicator do not char during sulfuric acid heat treatment; enhanced resistance to chloroform-methanol mixtures
  • Indicator: Contains fluorescent indicator (fluoresces under 254 nm radiation)
  • Convenience Features: Can be written on; easily cut to desired size allowing sample fractions to be removed directly

Specifications

Backing Material Plastic
Color White
Material Silica
Pore Size 120 Å
Quantity 50 Sheets
Type TLC Sheet
Fluorescent Indicator F254
For Use With (Application) TLC
Sorbent Silica Gel IB2
Thickness (Metric) 250 μm
Dimensions (L x W) 5 x 20 cm
Length (Metric) 20 cm
Width (Metric) 20 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.