Learn More
J.T. Baker™ BAKERBOND™ PROchievA™ Recombinant Protein A Resin, Affinity Chromatography Columns, Avantor
Catalog No. 02004668 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 x 1 mL Box
5 x 1 mL Box
5 x 5 mL Box
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-004-668 5 x 5 mL Box
02-004-665 1 x 1 mL Box
02-004-666 5 x 1 mL Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-004-668 Supplier J.T. Baker™ Supplier No. C78925
J.T.BAKER™ BAKERBOND™ PROchievA™ Recombinant Protein A Resin, Affinity Chromatography Columns offer excellent dynamic binding capacity and alkaline stability linked to Avantor's proprietary ligand.
  • The new protein A resin is designed for high-performance in the critical affinity chromatography step used in the purification of mAbs, Fc fusion proteins, and other similar IgG-like molecules.
  • Designed and manufactured by Avantor to the high standards established for J.T. Baker™ brand chemicals, BAKERBOND™ PROchievA™ recombinant protein
  • Resin can be used with other J.T. Baker™ BAKERBOND™ resins to enhance the separation power of chromatography process steps
  • Available in 1 mL and 5 mL lab columns designed for direct connection to many popular chromatography instruments including GE ÄKTA™ and Bio-Rad NGC™
Specifications
Product Type Affinity Chromatography Column
Binding Capacity FIO (Target >65 mg/mL)
Content And Storage 2°C to 8°C
Quantity 5 x 5 mL Box