Learn More
Avantor™ BAKERBOND™ spe Florisil Disposable Extraction Column
Catalog No. 02002439 Shop All Avantor Products
Change view
Click to view available options
Bed Weight:
500 mg
Catalog No. Bed Weight
02-002-439 500 mg
1 options
Catalog No. 02-002-439 Supplier Avantor™ Supplier No. 721306
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Define and control critical surface chemistry parameters to ensure performance consistency. Avantor BAKERBOND spe Florisil Disposable Extraction Columns, are designed with standard geometries to adapt to most popular automated liquid handling systems for extracting low to moderately polar analytes from nonaqueous solutions.
  • Constructed of ultraclean polypropylene
  • Provide recoveries when extracting analytes of weak hydrophobicity
  • Consistent bonding chemistry
  • Wide range of formats
Specifications
Column Capacity (Metric) 6 mL
Description Quantity: 30/Pk.
Bed Weight 500 mg
For Use With (Application) Low to Moderately Polar Analytes From Non-Aqueous Solutions
For Use With (Equipment) BAKER SPE-12 G Column Processor Complete (6998-00) or BAKER SPE-24 G Column Processor Complete (7208
Sorbent Florisil
Type Solid Phase Extraction Column