Learn More
Avantor™ BAKERBOND Speedisk™ H2O-Phobic DVB Extraction Disk
Catalog No. 02002350 Shop All Avantor Products
Change view
Click to view available options
Quantity:
20/Pk.
Catalog No. Quantity
02-002-350 20/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 02-002-350 Supplier Avantor™ Supplier No. 806806
This item is not returnable. View return policy
Maximize capacity, adsorption and SPE speed. Avantor BAKERBOND Speedisk Extraction Disk H2O-Phobic DVB are neither cartridge nor membrane that provides an excellent SPE tool for environmental samples and combines a thin bed of BAKERBOND sorbent with a special laminar configuration for testing drinking water, ground water, waste water, solid waste or process streams.
  • Pre-assembled with rigid support and baffle to maximize laminar flow, allow for filtration, resist clogging, and maximize capacity, adsorption, and speed
  • Require minimal glassware, set up time, and assembly
  • Minimizes impact of extractable compounds
  • Resists clogging and ensures high throughput rate
  • Reduce SPE run times
Specifications
For Use With (Application) Chlorinated Acids, EPA Method 515.2. Slightly Polar
For Use With (Equipment) Expanded Extraction Station (8095-06)
Type H2O-Phobic DVB Extraction Disk
Quantity 20/Pk.
Diameter (Metric) Outer 50 mm