Learn More
Avantor™ BAKERBOND Speedisk™ Octadecyl (C18) Extraction Disk
Catalog No. p-6827006 Shop All Avantor Products
Change view
Click to view available options
Sorbent:
C18
C18 (high capacity)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sorbent
02-002-375 C18
02-002-376 C18 (high capacity)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 02-002-375 Supplier Avantor™ Supplier No. 805506
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Maximize capacity, adsorption and SPE speed. Avantor BAKERBOND Speedisk Octadecyl (C18) Extraction Disks are neither cartridge nor membrane that provides an excellent SPE tool for environmental samples and combines a thin bed of BAKERBOND sorbent with a special laminar configuration for testing drinking water, ground water, waste water, solid waste or process streams.
  • Pre-assembled with rigid support and baffle to maximize laminar flow, allow for filtration, resist clogging, and maximize capacity, adsorption, and speed
  • Require minimal glassware, set up time, and assembly
  • Minimizes impact of extractable compounds
  • Resists clogging and ensures high throughput rate
  • Reduce SPE run times
Specifications
For Use With (Application) EPA Methods 500 Series, 608 SW 846/3535 and with Slightly Polar to Non-Polar Industrial Samples
For Use With (Equipment) Expanded Extraction Station (8095-06)
Sorbent C18
Type Extraction Disk
Quantity 20/Pk.
Diameter (Metric) Outer 50 mm