Learn More
Avantor™ BAKERBOND Speedisk™ Oil and Grease Extraction Disk
Catalog No. 02002379 Shop All Avantor Products
Change view
Click to view available options
Quantity:
20/Pk.
Catalog No. Quantity
02-002-379 20/Pk.
1 options
Catalog No. 02-002-379 Supplier Avantor™ Supplier No. 806006
Add to Cart
Add to Cart
Maximize capacity, adsorption and SPE speed. Avantor BAKERBOND Speedisk Oil and Grease Extraction Disk are neither cartridge nor membrane that provides an excellent SPE tool for environmental samples and combines a thin bed of BAKERBOND sorbent with a special laminar configuration for testing drinking water, ground water, waste water, solid waste or process streams.
  • Pre-assembled with rigid support and baffle to maximize laminar flow, allow for filtration, resist clogging, and maximize capacity, adsorption, and speed
  • Require minimal glassware, set up time, and assembly
  • Minimizes impact of extractable compounds
  • Resists clogging and ensures high throughput rate
  • Reduce SPE run times
Specifications
For Use With (Application) For Use with Slightly Polar to Non-Polar Hydrocarbons, EPA Method 1664, Rev. A.
For Use With (Equipment) Expanded Extraction Station (8095-06)
Type Oil and Grease Extraction Disk
Quantity 20/Pk.
Diameter (Metric) Outer 50 mm