Learn More
Bal Supply Cell Counters
Catalog No. p-139881 Shop All Tip Industries Inc. Products
Change view
Click to view available options
Style:
1 Counting unit; 1 reset knob
2 Counting units; 1 reset knob
5 Counting units; 2 reset knob
8 Counting units; 2 reset knob
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style
02-670-11 1 Counting unit; 1 reset knob
02-670-12 2 Counting units; 1 reset knob
02-670-13 5 Counting units; 2 reset knob
02-670-14 8 Counting units; 2 reset knob
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-670-11 Supplier Bal Supply Supplier No. 101C
Only null left
Add to Cart
Add to Cart
For automatic calculation including percentages.
For automatic calculation including percentages
  • Totaling unit rings a bell at 100 increments
  • Dual reset knobs
  • Last unit is the totaling unit, which records total number of strokes made on other keys
  • Removable tabs on top of counter provide area for identification of classification being counted
Specifications
Product Type Differential Cell Counter
Displayed Text 0 to 999
Style 1 Counting unit; 1 reset knob
For Use With (Application) Cell counting with automatic calculations