Learn More
Bal Supply Slotted Lattice Connector
Catalog No. 13684136 Shop All Tip Industries Inc. Products
Change view
Click to view available options
Size:
0.5 in.
Catalog No. Size
13-684-136 0.5 in.
1 options
Catalog No. 13-684-136 Supplier Bal Supply Supplier No. 4010LC
Add to Cart
Add to Cart
Includes
Allen wrench
Made of cast aluminum with a smooth round surface for connecting lattice rod supports for laboratory apparatus
  • Spherical designed connectors assure rigidity, solidity and freedom from breakage
  • U-shaped slots on each side take standard 1/2-inch rod and make possible many combinations of rod supports
  • Axial hole permits rods to be jointed at right angles in the same plane
  • Open U slot allows removal or addition of rods to a laboratory setup without disturbing the whole structure
  • Set screws are easily tightened with the allen wrench
  • Screws, when tightened in place, are flush, eliminating the inconvenience of protruding locking devices
  • 12/case
Specifications
Material Cast Aluminum
Includes U-shape slots
Product Type Lattice Connector
Size 0.5 in.
For Use With (Application) Connecting lattice rod supports for laboratory apparatus
For Use With (Equipment) 0.5 in. Rod