Learn More
Bal Supply Stainless-Steel Pinch Clamps for Ball and Socket Joints
Catalog No. 13684139 Shop All Tip Industries Inc. Products
Change view
Click to view available options
Size:
12
18
28
35
75
Quantity:
12/Pk.
6/Pk.
Includes:
No Screw Locking Device
Screw Locking Device
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Quantity Includes
13-684-139 75 6/Pk. Screw Locking Device
05-885B 12 12/Pk. No Screw Locking Device
05-885D 18 12/Pk. No Screw Locking Device
05-885F 28 12/Pk. Screw Locking Device
05-885C 12 12/Pk. Screw Locking Device
05-885E 18 12/Pk. Screw Locking Device
05-885G 35 12/Pk. Screw Locking Device
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 13-684-139 Supplier Bal Supply Supplier No. 20275WS
Add to Cart
Add to Cart
Easy, secure way to marry ball-and-socket joints
Spring-loaded pinch clamp with forked jaws on both sides of the joint ensures tight fit.
  • 05-885C and 05-885E through K have screw-locking device
  • Work well on both O-ring and ball-and-socket joints
  • Others are intended for ball-and-socket joints only
Specifications
Quantity 6/Pk.
Includes Screw Locking Device
Size 75