Learn More
Medchemexpress LLC HY-P1031 1mg Medchemexpress, [bAla8]-Neurokinin A(4-10) CAS:122063-01-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP10311MG
Encompass_Preferred
Medchemexpress, HY-P1031 1mg [bAla8]-Neurokinin A(4-10) CAS:122063-01-8 [bAla8]-Neurokinin A(4-10) is a neurokinin 2 (NK2) receptor agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4140