Learn More
Grainger Balaclava Black Fleece
Catalog No. 502764395
Encompass_Preferred
The N-FERNO Flame Resistant Balaclava by Ergodyne is a flame resistant balaclava made from Nomex and Kevlar blend fabric. It features a single layer design that provides protection from flash fires and electrical arc flash. The balaclava has a pull-on design with a 3-panel hood for full head and neck coverage. It meets NFPA 70E and 2112 standards for flame resistant clothing. The balaclava comes in black color in one universal size that fits most. It is lightweight, breathable, and moisture-wicking. This balaclava provides protection for welders, utility workers, and other professionals working in high heat environments.
Catalog No. 50-276-4395 Supplier Grainger Supplier No. 16W658
The N-FERNO Flame Resistant Balaclava by Ergodyne is a flame resistant balaclava made from Nomex and Kevlar blend fabric. It features a single layer design that provides protection from flash fires and electrical arc flash. The balaclava has a pull-on design with a 3-panel hood for full head and neck coverage. It meets NFPA 70E and 2112 standards for flame resistant clothing. The balaclava comes in black color in one universal size that fits most. It is lightweight, breathable, and moisture-wicking. This balaclava provides protection for welders, utility workers, and other professionals working in high heat environments.