Learn More
FUJIFILM Biosciences BalanCD CHO Feed 1 - Liquid
Catalog No. NC0373281
Encompass
BalanCD CHO Feed 1 is a chemically-defined, animal component-free feed medium designed to increase process yields of antibodies and recombinant proteins in Chinese Hamster Ovary (CHO) cells in fed-batch mode. This formulation was developed using FUJIFILM Irvine Scientific’s Rational Culture Media Design strategy to achieve enhanced performance of growth and production in fed-batch culture.BalanCD CHO Feed 1 was designed to be used in conjunction with BalanCD CHO Growth A and can work with a wide range of growth media for optimal performance in fed-batch cultures.
Catalog No. NC0373281 Supplier FUJIFILM Biosciences Supplier No. 911271L
BalanCD CHO Feed 1 is a chemically-defined, animal component-free feed medium designed to increase process yields of antibodies and recombinant proteins in Chinese Hamster Ovary (CHO) cells in fed-batch mode. This formulation was developed using FUJIFILM Irvine Scientific’s Rational Culture Media Design strategy to achieve enhanced performance of growth and production in fed-batch culture.BalanCD CHO Feed 1 was designed to be used in conjunction with BalanCD CHO Growth A and can work with a wide range of growth media for optimal performance in fed-batch cultures.