Learn More
FUJIFILM Biosciences BalanCD CHO Feed 1 - Powder
Catalog No. 5000450186
Encompass
BalanCD CHO Feed 1 is a chemically-defined animal component-free feed medium designed to increase process yields of antibodies and recombinant proteins in Chinese Hamster Ovary CHO cells in fed-batch mode This formulation was developed using FUJIFILM Irvine Scientific s Rational Culture Media Design strategy to achieve enhanced performance of growth and production in fed-batch culture
Catalog No. 50-004-50186 Supplier FUJIFILM Biosciences Supplier No. 9411910L
BalanCD CHO Feed 1 is a chemically-defined animal component-free feed medium designed to increase process yields of antibodies and recombinant proteins in Chinese Hamster Ovary CHO cells in fed-batch mode This formulation was developed using FUJIFILM Irvine Scientific s Rational Culture Media Design strategy to achieve enhanced performance of growth and production in fed-batch culture