Learn More
FUJIFILM Biosciences BalanCD CHO Perfusion - Liquid
Catalog No. 5000450301
Encompass
BalanCD CHO Perfusion is a chemically defined medium that is optimized for the growth and production of suspension CHO cell cultures in continuous steady-state and N-1 perfusion cultures As a high performing medium it can achieve key perfusion metrics including cell-specific productivity volumetric productivity and cell-specific perfusion rate CSPR BalanCD CHO Perfusion medium meets the requirements ofLarge scale industrial cell cultures and is intended forLong-term continuous culturing of CHO cells
Catalog No. 50-004-50301 Supplier FUJIFILM Biosciences Supplier No. 911781L
BalanCD CHO Perfusion is a chemically defined medium that is optimized for the growth and production of suspension CHO cell cultures in continuous steady-state and N-1 perfusion cultures As a high performing medium it can achieve key perfusion metrics including cell-specific productivity volumetric productivity and cell-specific perfusion rate CSPR BalanCD CHO Perfusion medium meets the requirements ofLarge scale industrial cell cultures and is intended forLong-term continuous culturing of CHO cells