Learn More
FUJIFILM Biosciences BalanCD CHO Perfusion - Powder
Catalog No. 5000450187
Encompass
Support the growth and productivity of CHO cells in perfusion culture systems with BalanCD CHO Perfusion a chemically defined complete and ready-to-use medium Optimize and achieve key perfusion metrics including cell-specific productivity volumetric productivity and cell-specific perfusion rate CSPR for continuous steady-state and N-1 perfusion cultures
Catalog No. 50-004-50187 Supplier FUJIFILM Biosciences Supplier No. 9414910L
Support the growth and productivity of CHO cells in perfusion culture systems with BalanCD CHO Perfusion a chemically defined complete and ready-to-use medium Optimize and achieve key perfusion metrics including cell-specific productivity volumetric productivity and cell-specific perfusion rate CSPR for continuous steady-state and N-1 perfusion cultures