Learn More
FUJIFILM Biosciences BalanCD HEK293 - Liquid
Catalog No. NC1192689
Encompass
BalanCD HEK293 medium is a versatile, chemically-defined, animal component-free medium optimized for transfection and production of viral vectors and recombinant proteins.
Catalog No. NC1192689 Supplier FUJIFILM Biosciences Supplier No. 911651L
BalanCD HEK293 medium is a versatile, chemically-defined, animal component-free medium optimized for transfection and production of viral vectors and recombinant proteins.