Learn More
Scientific Industries BALANCE, ANALYTICAL, 220G
Catalog No. 501139357
Encompass_Preferred
Analytical Balance, 220g x 0.0001g, offers superb precision and accuracy. The AGZN is built with a die-cast metal housing and large easily accessible glass weighing chamber. The balance includes 5 weighing functions including density calculation,and equipped with RS232, USB, Clock, Backlit LCD Display.
Catalog No. 50-113-9357 Supplier Scientific Industries Supplier No. AGZN220
Analytical Balance, 220g x 0.0001g, offers superb precision and accuracy. The AGZN is built with a die-cast metal housing and large easily accessible glass weighing chamber. The balance includes 5 weighing functions including density calculation,and equipped with RS232, USB, Clock, Backlit LCD Display.