Learn More
METTLER TOLEDO 3P INDUSTRIAL Balance Protective Cover MS-L
Catalog No. NC1802944
Encompass_Preferred
The protective cover decreases the need to clean your balance and helps prevents cross contamination. Simply replace as required. The chemically resistant cover protects the balance against toxic or hazardous substances and provides operators with an increased level of safety. The cover is installed and removed in a few simple steps. Once in place, it does not interfere with balance operation.
Catalog No. NC1802944 Supplier METTLER TOLEDO 3P INDUSTRIAL Supplier No. 12121852
The protective cover decreases the need to clean your balance and helps prevents cross contamination. Simply replace as required. The chemically resistant cover protects the balance against toxic or hazardous substances and provides operators with an increased level of safety. The cover is installed and removed in a few simple steps. Once in place, it does not interfere with balance operation.