Learn More
GSC Go Science Crazy Balance Demonstration Stand
SCI_ED
Catalog No. S24615 Shop All GSC Products
Encompass
Encompass_Preferred
Catalog No. S24615 Supplier GSC Go Science Crazy Supplier No. 415001
Force and Motion Demonstration
The Balance Demonstration Apparatus includes the fulcrum with metal support base and upright aluminum support arm, one meter stick and one knife edge clamp.
Specifications
Product Type New York Balance
For Use With (Application) Principles and properties of torques
Includes Measuring stick, 50 cm; Fulcrum; Knife edge clamp
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov